8-Bromchinolin-2(1H)-on

Lieferant: COMBI-BLOCKS

PY-1183-5G PY-1183-1G
COBBPY-1183-5GEA 543 EUR
COBBPY-1183-5G COBBPY-1183-1G
8-Bromchinolin-2(1H)-on
8-Bromchinolin-2(1H)-on
Formel: C₉H₆BrNO
Molecular Weight: 224,0540 g/mol
MDL: MFCD08741409
CAS-Nummer: 67805-67-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR