4-Methylsalicylaldehyd, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Hydroxy-4-methylbenzaldehyde
M-Clarity™ Qualitätsstufe = MQ300
W369705-100G-K
W369705-25G-K
SIALW369705-100G-KEA
1380
EUR
SIALW369705-100G-K
SIALW369705-25G-K
4-Methylsalicylaldehyd, Sigma-Aldrich®
4-Methylsalicylaldehyd
Formel:
C₈H₈O₂ Molecular Weight: 136,15 g/mol |
MDL:
MFCD00799550 CAS-Nummer: 698-27-1 EINECS: 211-813-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...