Chlordiisopropyloctylsilan
Lieferant: CHEMPUR SILANE
SILS12610.1
SILS12610.10
SILS12610.25
SILNSILS12610.1EA
0
EUR
SILNSILS12610.1
SILNSILS12610.10
SILNSILS12610.25
Chlordiisopropyloctylsilan
Chlordiisopropyloctylsilan
Formel:
C₁₄H₃₁ClSi |
MDL:
MFCD00134528 CAS-Nummer: 117559-37-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...