Chlordimethyloctylsilan
Lieferant: CHEMPUR SILANE
SILS12625.10
SILS12625.25
SILS12625.100
SILNSILS12625.10EA
0
EUR
SILNSILS12625.10
SILNSILS12625.25
SILNSILS12625.100
Chlordimethyloctylsilan
Chlordimethyloctylsilan
Formel:
C₁₀H₂₃ClSi |
MDL:
MFCD00000511 CAS-Nummer: 18162-84-0 EINECS: 242-044-5 UN: 2987 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...