Isofluran ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 97%
CAS No.: 26675-46-7
Molecular Formula: CF3CHClOCHF2
Formula Weight: 184.49
Density: 1.496
Refractive Index:
Formel:
C₃H₂ClF₅O Siedepunkt: 48…49 °C Dichte: 1,496 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066609 CAS-Nummer: 26675-46-7 EINECS: 247-897-7 Merck Index: 14,5175 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...