1-Butyl-4-piperidon

Lieferant: COMBI-BLOCKS

Synonyme: 1-Butylpiperidin-4-one

HC-6269-5G HC-6269-25G HC-6269-1G
COBBHC-6269-5GEA 404 EUR
COBBHC-6269-5G COBBHC-6269-25G COBBHC-6269-1G
1-Butyl-4-piperidon
1-Butyl-4-piperidon
Formel: C₉H₁₇NO
Molecular Weight: 155,24 g/mol
MDL: MFCD00023404
CAS-Nummer: 23081-86-9
EINECS: 245-420-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR