4-tert-Butylbenzophenon ≥98%

Lieferant: Thermo Scientific

L06386.03 L06386.06
L06386.03EA 43.3 EUR
L06386.03 L06386.06
4-tert-Butylbenzophenon ≥98%
4-tert-Butylbenzophenon

Purity: 98% CAS No.: 22679-54-5 Molecular Formula: C17H18O Formula Weight: 238.32 Flash Point: 150°(302°F) Melting Point: 38-42° Boiling Point: 168-170°/2mm Beilstein Reference: 2100075 MDL No.: MFCD00017255

Formel: C₁₇H₁₈O
Molecular Weight: 238,32 g/mol
Siedepunkt: 168…170 °C (2 mmHg)
Schmelzpunkt: 38…42 °C
Flammpunkt: 150 °C (302 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00017255
CAS-Nummer: 22679-54-5
EINECS: 245-154-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR