Toluol-[Ring-D5] (98% D)
Lieferant: LGC Standards PROMOCHEM
|
Gefahr
|
Synonyme:
Toluene-[ring-²H₅]
CIL-DLM-1176-1
CIL-DLM-1176-5
PROOCIL-DLM-1176-1EA
263
EUR
PROOCIL-DLM-1176-1
PROOCIL-DLM-1176-5
Toluol-[Ring-D5] (98% D)
Toluol-[Ring-D5]
Formel:
C₆D₅CH₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00084186 CAS-Nummer: 1603-99-2 UN: 1294 ADR: 3,II |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...