4-Propoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-n-Propoxybenzeneboronic acid

BB-2075-1G BB-2075-25G BB-2075-5G
COBBBB-2075-1GEA 35.1 EUR
COBBBB-2075-1G COBBBB-2075-25G COBBBB-2075-5G
4-Propoxyphenylboronsäure
4-Propoxyphenylboronsäure
Formel: C₉H₁₃BO₃
Molecular Weight: 180.01 g/mol
MDL: MFCD03427189
CAS-Nummer: 186497-67-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR