5-Brom-m-kresol

Lieferant: COMBI-BLOCKS

Synonyme: 3-Bromo-5-methylphenol

OT-1236-5G OT-1236-25G OT-1236-1G
COBBOT-1236-5GEA 157 EUR
COBBOT-1236-5G COBBOT-1236-25G COBBOT-1236-1G
5-Brom-m-kresol
5-Brom-m-kresol
Formel: C₇H₇BrO
Molecular Weight: 187,0300 g/mol
MDL: MFCD09038281
CAS-Nummer: 74204-00-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR