Docosyltrichlorsilan

Lieferant: CHEMPUR SILANE

SILS08475.5 SILS08475.25
SILNSILS08475.5EA 0 EUR
SILNSILS08475.5 SILNSILS08475.25
Docosyltrichlorsilan
Docosyltrichlorosilane
Formel: C₂₂H₄₅Cl₃Si
CAS-Nummer: 7325-84-0
EINECS: 230-802-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR