4-(Chlormethyl)phenyltrichlorsilan

Lieferant: CHEMPUR SILANE

SILS04325.5 SILS04325.25
SILNSILS04325.5EA 0 EUR
SILNSILS04325.5 SILNSILS04325.25
4-(Chlormethyl)phenyltrichlorsilan
4-(Chlormethyl)phenyltrichlorsilan
Formel: C₇H₆Cl₄Si
Molecular Weight: 260.02 g/mol
MDL: MFCD00045268
CAS-Nummer: 13688-90-9
EINECS: 237-203-0
UN: 2987
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR