2,6-Dichlorisonicotinamid

Lieferant: Thermo Scientific

RF01160.250 RF01160.1
MAYBRF01160.250EA 0 EUR
MAYBRF01160.250 MAYBRF01160.1
2,6-Dichlorisonicotinamid
2,6-Dichlorisonicotinamid
Formel: C₆H₄Cl₂N₂O
Molecular Weight: 191.0200 g/mol
MDL: MFCD00052640
CAS-Nummer: 89281-13-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR