2-Amino-4-chlor-3,5,6-trifluorpyridin 98%
Lieferant: Thermo Scientific
|
Achtung
|
H34155.03
H34155.06
H34155.03EA
80.3
EUR
H34155.03
H34155.06
2-Amino-4-chlor-3,5,6-trifluorpyridin 98%
2-Amino-4-chloro-3,5,6-trifluoropyridine
Purity: 98%
CAS No.: 63489-56-5
Molecular Formula: C5H2ClF3N2
Formula Weight: 182.54
Melting Point: 119-123°
MDL No.: MFCD08436088
Formel:
C₅H₂ClF₃N₂ Molecular Weight: 182,54 g/mol Schmelzpunkt: 119…123 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08436088 CAS-Nummer: 63489-56-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...