4-Amino-2,3,5,6-tetrafluorpyridin

Lieferant: FLUOROCHEM

Synonyme: 2,3,5,6-Tetrafluoropyridin-4-amine

005234-1G 005234-5G 005234-25G
FLUO005234-1GEA 31.1 EUR
FLUO005234-1G FLUO005234-5G FLUO005234-25G
4-Amino-2,3,5,6-tetrafluorpyridin
4-Amino-2,3,5,6-tetrafluoropyridine
Formel: C₅H₂F₄N₂
Molecular Weight: 166.08 g/mol
MDL: MFCD00010855
CAS-Nummer: 1682-20-8

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR