2-Amino-3,6-difluor-4-(trifluormethyl)pyridin

Lieferant: FLUOROCHEM

013111-250MG 013111-1G
FLUO013111-250MGEA 0 EUR
FLUO013111-250MG FLUO013111-1G
2-Amino-3,6-difluor-4-(trifluormethyl)pyridin
2-Amino-3,6-difluoro-4-(trifluoromethyl)pyridine
Formel: C₆H₃F₅N₂
MDL: MFCD03840200
CAS-Nummer: 675602-89-8
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR