2-Amino-3-fluor-5-(trifluormethyl)pyridin 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
3-Fluoro-5-(trifluoromethyl)pyridin-2-amine
H32532.03
H32532.06
H32532.03EA
66.1
EUR
H32532.03
H32532.06
2-Amino-3-fluor-5-(trifluormethyl)pyridin 97%
2-Amino-3-fluoro-5-(trifluoromethyl)pyridine
Purity: 97%
CAS No.: 852062-17-0
Molecular Formula: C6H4F4N2
Formula Weight: 180.10
Melting Point: 64-68°
UN No.: 2811
MDL No.: MFCD10699117
Formel:
C₆H₄F₄N₂ Molecular Weight: 180,10 g/mol Schmelzpunkt: 64…68 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD10699117 CAS-Nummer: 852062-17-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...