D(+)-Cellobiose 98%

Lieferant: Thermo Scientific

Synonyme: β-D-Glc-(1→4)-D-Glc, 4-β-D-glucopyranosyl-D-glueopyranose, Cellose, β-Cellobiose, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, 4-O-β-D-Glucopyranosyl-D-glucopyranose

108460050 108460250 108461000 108465000
ACRO108460050EA 39.4 EUR
ACRO108460050 ACRO108460250 ACRO108461000 ACRO108465000
D(+)-Cellobiose 98%
D(+)-Cellobiose
Formel: C₁₂H₂₂O₁₁
Molecular Weight: 342,30 g/mol
Schmelzpunkt: 239 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00136034
CAS-Nummer: 528-50-7
EINECS: 208-436-5

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Solubility (5 % in water) Clear colorless
HPLC ≥97.5 % (ELSD)
Specific optical rotation +33° to +35° (20°C, 589 nm) (c=1, water) (after 15 h)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR