D(+)-Cellobiose, Millipore®

Lieferant: Merck

Synonyme: β-D-Glc-(1→4)-D-Glc, 4-β-D-glucopyranosyl-D-glueopyranose, Cellose, β-Cellobiose, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, 4-O-β-D-Glucopyranosyl-D-glucopyranose

M-Clarity™ Qualitätsstufe = MQ100

1.02352.0025 1.02352.1000
1.02352.0025EA 222 EUR
1.02352.0025 1.02352.1000
D(+)-Cellobiose, Millipore®
D(+)-Cellobiose
Formel: C₁₂H₂₂O₁₁
Molecular Weight: 342,30 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00136034
CAS-Nummer: 528-50-7
EINECS: 208-436-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR