D(+)-Turanose, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(3S,4R,5R)-1,4,5,6-Tetrahydroxy-3-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexan-2-one, Turanose
M-Clarity™ Qualitätsstufe = MQ200
T2754-1G
T2754-25G
T2754-5G
SIALT2754-1GEA
72
EUR
SIALT2754-1G
SIALT2754-25G
SIALT2754-5G
D(+)-Turanose, Sigma-Aldrich®
D(+)-Turanose
Formel:
C₁₂H₂₂O₁₁ Molecular Weight: 342.30 g/mol |
MDL:
MFCD00006606 CAS-Nummer: 547-25-1 EINECS: 208-918-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...