Kaliumbis(trimethylsilyl)amid 12% in Toluol

Lieferant: CHEMPUR SILANE

SILS09635.25 SILS09635.100
SILNSILS09635.25EA 0 EUR
SILNSILS09635.25 SILNSILS09635.100
Kaliumbis(trimethylsilyl)amid 12% in Toluol
Kaliumbis(trimethylsilyl)amid
Formel: C₆H₁₈KNSi₂
MDL: MFCD00010330
CAS-Nummer: 40949-94-8
EINECS: 424-100-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR