Kaliumbis(trimethylsilyl)amid 20% in THF
Lieferant: CHEMPUR SILANE
SILS13950.25
SILS13950.100
SILNSILS13950.25EA
0
EUR
SILNSILS13950.25
SILNSILS13950.100
Kaliumbis(trimethylsilyl)amid 20% in THF
Kaliumbis(trimethylsilyl)amid
Formel:
C₆H₁₈KNSi₂ |
MDL:
MFCD00010330 CAS-Nummer: 40949-94-8 EINECS: 424-100-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...