Natriumbis(trimethylsilyl)amid 30% in THF

Lieferant: CHEMPUR SILANE

Synonyme: Natriumbis(trimethylsilyl)amid, HMDS Natriumsalz, N-Natriohexamethyldisilazan, HMDS-Na

SILS14980.100 SILS14980.250
SILNSILS14980.100EA 0 EUR
SILNSILS14980.100 SILNSILS14980.250
Natriumbis(trimethylsilyl)amid 30% in THF
Natriumbis(trimethylsilyl)amid
Formel: C₆H₁₈NNaSi₂
Molecular Weight: 183,38 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00009835
CAS-Nummer: 1070-89-9
EINECS: 213-983-8
UN: 2924
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR