2'-Brom-5'-chlor-4'-methylacetanilid

Lieferant: COMBI-BLOCKS

OT-0066-5G OT-0066-1G
COBBOT-0066-5GEA 98.3 EUR
COBBOT-0066-5G COBBOT-0066-1G
2'-Brom-5'-chlor-4'-methylacetanilid
2'-Brom-5'-chlor-4'-methylacetanilid
Formel: C₉H₉BrClNO
MDL: MFCD12546456
CAS-Nummer: 116010-06-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR