Ethylchlorfluoracetat

Lieferant: FLUOROCHEM

003216-5G 003216-25G 003216-100G
FLUO003216-5GEA 0 EUR
FLUO003216-5G FLUO003216-25G FLUO003216-100G
Ethylchlorfluoracetat
Ethylchlorfluoracetat
Formel: C₄H₆ClFO₂
MDL: MFCD00039333
CAS-Nummer: 401-56-9
EINECS: 206-930-5
UN: 2920
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR