Ethylchlorfluoracetat
Lieferant: CHEMPUR
Synonyme:
Ethylchlorofluoroacetate
BTB11157.250
BTB11157.1
CHMPBTB11157.250EA
0
EUR
CHMPBTB11157.250
MAYBBTB11157.1
Ethylchlorfluoracetat
Ethylchlorfluoracetat
Formel:
C₄H₆ClFO₂ Molecular Weight: 140,54 g/mol |
MDL:
MFCD00039333 CAS-Nummer: 401-56-9 EINECS: 206-930-5 UN: 2920 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...