2,2,2-Trifluorethylbutyrat

Lieferant: FLUOROCHEM

Synonyme: Butyric acid 2,2,2-Trifluoroethyl ester

007518-5G 007518-25G
FLUO007518-5GEA 32.3 EUR
FLUO007518-5G FLUO007518-25G
2,2,2-Trifluorethylbutyrat
2,2,2-Trifluorethylbutyrat
Formel: C₆H₉F₃O₂
Molecular Weight: 170,1297 g/mol
MDL: MFCD00077609
CAS-Nummer: 371-27-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR