2,2,2-Trifluorethylformiat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

669083-1G 669083-5G
SIAL669083-1GEA 30.2 EUR
SIAL669083-1G SIAL669083-5G
2,2,2-Trifluorethylformiat, Sigma-Aldrich®
2,2,2-Trifluorethylformiat
Formel: C₃H₃F₃O₂
Molecular Weight: 128.05 g/mol
MDL: MFCD06797733
CAS-Nummer: 32042-38-9
UN: 3272
ADR: 3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR