Methylpentafluorpropionat
Lieferant: FLUOROCHEM
Synonyme:
Methyl 2,2,3,3,3-pentafluoropropanoate
001207-25G
001207-100G
FLUO001207-25GEA
37.8
EUR
FLUO001207-25G
FLUO001207-100G
Methylpentafluorpropionat
Methylpentafluorpropionat
Formel:
C₄H₃F₅O₂ Molecular Weight: 178,06 g/mol |
MDL:
MFCD00000430 CAS-Nummer: 378-75-6 EINECS: 206-828-0 UN: 3272 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...