2,2,2-Trifluorethyltrifluormethansulfonat
Lieferant: FLUOROCHEM
Synonyme:
2,2,2-Trifluoroethyltrifluoromethanesulphonate
008628-5G
008628-25G
008628-1G
FLUO008628-5GEA
90.9
EUR
FLUO008628-5G
FLUO008628-25G
FLUO008628-1G
2,2,2-Trifluorethyltrifluormethansulfonat
2,2,2-Trifluorethyltrifluormethansulfonat
Formel:
C₃H₂F₆O₃S Molecular Weight: 232.10 g/mol |
MDL:
MFCD00671579 CAS-Nummer: 6226-25-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...