Ethylpentafluorbenzoat

Lieferant: FLUOROCHEM

Synonyme: Ethyl 2,3,4,5,6-pentafluorobenzoate

006123-5G 006123-25G 006123-100G
FLUO006123-5GEA 50.1 EUR
FLUO006123-5G FLUO006123-25G FLUO006123-100G
Ethylpentafluorbenzoat
Ethylpentafluorbenzoat
Formel: C₉H₅F₅O₂
Molecular Weight: 240.13 g/mol
MDL: MFCD00039211
CAS-Nummer: 4522-93-4
EINECS: 224-854-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR