3'-Brom-4'-chloracetophenon

Lieferant: COMBI-BLOCKS

Synonyme: 3-Bromo-4-chloroacetophenone

CA-4666-5G CA-4666-1G
COBBCA-4666-5GEA 692 EUR
COBBCA-4666-5G COBBCA-4666-1G
3'-Brom-4'-chloracetophenon
3'-Brom-4'-chloracetophenon
Formel: C₈H₆BrClO
MDL: MFCD09832932
CAS-Nummer: 54826-14-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR