ω,ω-Dibrom-4-fluoracetophenon ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2,2-Dibromo-4'-fluoroacetophenone
L19788.03
L19788.06
L19788.03EA
12
EUR
L19788.03
L19788.06
ω,ω-Dibrom-4-fluoracetophenon ≥98%
ω,ω-Dibrom-4-fluoracetophenon
Purity: 98%
CAS No.: 7542-64-5
Molecular Formula: C8H5Br2FO
Formula Weight: 295.93
Storage Temperature: 2°C to 8°C
Melting Point: 36-38°
Beilstein Reference: 2256317
UN No.: 3261
MDL No.: MFCD03791309
Formel:
C₈H₅Br₂FO Molecular Weight: 295.93 g/mol Schmelzpunkt: 36…38 °C |
MDL:
MFCD03791309 CAS-Nummer: 7542-64-5 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...