4'-Brom-2,2,2-trifluoracetophenon ≥97%

Lieferant: Thermo Scientific
Achtung

H33627.03 H33627.06
H33627.03EA 128 EUR
H33627.03 H33627.06
4'-Brom-2,2,2-trifluoracetophenon ≥97%
4'-Brom-2,2,2-trifluoracetophenon

Purity: 97% CAS No.: 16184-89-7 Molecular Formula: C8H4BrF3O Formula Weight: 253.02 Melting Point: 26-30° Boiling Point: 95°/4mm MDL No.: MFCD00191862

Formel: C₈H₄BrF₃O
Molecular Weight: 253,02 g/mol
Siedepunkt: 95 °C (4 mmHg)
Schmelzpunkt: 26…30 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00191862
CAS-Nummer: 16184-89-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR