4'-Brom-2,2,2-trifluoracetophenon, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
403229-1G
403229-5G
SIAL403229-1GEA
21.9
EUR
SIAL403229-1G
SIAL403229-5G
4'-Brom-2,2,2-trifluoracetophenon, Sigma-Aldrich®
4'-Brom-2,2,2-trifluoracetophenon
Formel:
C₈H₄BrF₃O Molecular Weight: 253,02 g/mol |
MDL:
MFCD00191862 CAS-Nummer: 16184-89-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...