Antimon(III)isopropoxid ≥99,9% (Metall-Basis)

Lieferant: Thermo Scientific
Achtung

36589.06 36589.14
36589.06EA 116 EUR
36589.06 36589.14
Antimon(III)isopropoxid ≥99,9% (Metall-Basis)
Antimon(III)isopropoxid

Purity: 99.9% CAS No.: 18770-47-3 Molecular Formula: Sb[OCH(CH3)2]3 Formula Weight: 298.80 Flash Point: 77°(141°F) Physical Form: Liquid Density: 1.31 Sensitivity: Moisture Sensitive Appearance: Colourless Boiling Point: 68°/3mm UN No.: 3082 MDL No.: MFCD00143623

Formel: C₉H₂₁O₃Sb
Molecular Weight: 298,80 g/mol
Siedepunkt: 68 °C (3 mmHg)
Dichte: 1,31
Flammpunkt: 77 °C (141 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00143623
CAS-Nummer: 18770-47-3
EINECS: 242-561-6
UN: 3082
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR