Trimethylamin-N-oxid Dihydrat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Trimethylamine oxide dihydrate
M-Clarity™ Qualitätsstufe = MQ200
92277-10G
92277-50G
SIAL92277-10GEA
47.9
EUR
SIAL92277-10G
SIAL92277-50G
Trimethylamin-N-oxid Dihydrat, Sigma-Aldrich®
Trimethylamin-N-oxid Dihydrat
Formel:
C₃H₉NO·2H₂O Molecular Weight: 111,14 g/mol (75,11 anhy) |
MDL:
MFCD00149077 CAS-Nummer: 62637-93-8 EINECS: 214-675-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...