Geranylacetat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(E)-3,7-Dimethylocta-2,6-dien-1-yl acetate
M-Clarity™ Qualitätsstufe = MQ100
173495-100G
173495-25G
SIAL173495-100GEA
89.1
EUR
SIAL173495-100G
SIAL173495-25G
Geranylacetat, Sigma-Aldrich®
Geranylacetat
Formel:
C₁₂H₂₀O₂ Molecular Weight: 196,29 g/mol |
MDL:
MFCD00015037 CAS-Nummer: 105-87-3 EINECS: 203-341-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...