5-Brom-4-methyl-2-nitroanilin
Lieferant: COMBI-BLOCKS
HC-2908-5G
HC-2908-1G
HC-2908-250M
COBBHC-2908-5GEA
712
EUR
COBBHC-2908-5G
COBBHC-2908-1G
COBBHC-2908-250M
5-Brom-4-methyl-2-nitroanilin
5-Brom-4-methyl-2-nitroanilin
Formel:
C₇H₇BrN₂O₂ Molecular Weight: 231,0500 g/mol |
MDL:
MFCD00156314 CAS-Nummer: 40371-63-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...