5-Chlor-2-nitro-p-toluidin
Lieferant: COMBI-BLOCKS
|
Gefahr
|
Synonyme:
5-Chloro-4-methyl-2-nitroaniline
HC-2909-1G
HC-2909-25G
HC-2909-5G
COBBHC-2909-1GEA
0
EUR
COBBHC-2909-1G
COBBHC-2909-25G
COBBHC-2909-5G
5-Chlor-2-nitro-p-toluidin
5-Chlor-2-nitro-p-toluidin
Formel:
C₇H₇ClN₂O₂ |
MDL:
MFCD00034066 CAS-Nummer: 7149-80-6 UN: 2237 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...