1-Mesityl-2-thioharnstoff

Lieferant: Thermo Scientific

Synonyme: N-(2,4,6-Trimethylphenyl)thiourea

S03098.1 S03098.50
MAYBS03098.1EA 0 EUR
MAYBS03098.1 MAYBS03098.50
1-Mesityl-2-thioharnstoff
1-Mesityl-2-thioharnstoff
Formel: C₁₀H₁₄N₂S
Molecular Weight: 194,30 g/mol
MDL: MFCD00051488
CAS-Nummer: 91059-97-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR