1-[3,5-Bis(trifluoromethyl)phenyl]pyrrole

Lieferant: FLUOROCHEM

008212-1G 008212-5G
FLUO008212-1GEA 57.9 EUR
FLUO008212-1G FLUO008212-5G
1-[3,5-Bis(trifluoromethyl)phenyl]pyrrole
1-[3,5-Bis(trifluoromethyl)phenyl]pyrrole
Formel: C₁₂H₇F₆N
Molecular Weight: 279,19 g/mol
MDL: MFCD00053048
CAS-Nummer: 175136-60-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR