Tris[3,5-bis(trifluormethyl)phenyl]phosphin
Lieferant: FLUOROCHEM
Synonyme:
Tris(3,5-bis(trifluoromethyl)phenyl)phosphine
007746-1G
007746-5G
FLUO007746-1GEA
79.1
EUR
FLUO007746-1G
FLUO007746-5G
Tris[3,5-bis(trifluormethyl)phenyl]phosphin
Tris[3,5-bis(trifluormethyl)phenyl]phosphin
Formel:
C₂₄H₉F₁₈P Molecular Weight: 670.28 g/mol |
MDL:
MFCD00042428 CAS-Nummer: 175136-62-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...