4'-Benzyloxy-3'-fluoracetophenon
Lieferant: BIONET RESEARCH
Synonyme:
1-(4-(Benzyloxy)-3-fluorophenyl)ethanone
3X-0863.1
3X-0863.10
3X-0863.5
3X-0863.500
BIOT3X-0863.1EA
0
EUR
BIOT3X-0863.1
BIOT3X-0863.10
BIOT3X-0863.5
BIOT3X-0863.500
4'-Benzyloxy-3'-fluoracetophenon
4'-Benzyloxy-3'-fluoracetophenon
Formel:
C₁₅H₁₃FO₂ Molecular Weight: 244.261 g/mol |
MDL:
MFCD06659550 CAS-Nummer: 81227-99-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...