2-Methoxy-N-methyanilin

Lieferant: FLUOROCHEM

Synonyme: 2-Methoxy-N-methylaniline

009652-25G 009652-5G
FLUO009652-25GEA 0 EUR
FLUO009652-25G FLUO009652-5G
2-Methoxy-N-methyanilin
2-Methoxy-N-methyanilin
Formel: C₈H₁₁NO
Molecular Weight: 137.1790 g/mol
MDL: MFCD03093822
CAS-Nummer: 10541-78-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR