L(-)-Fucose 98%
Lieferant: Thermo Scientific
Synonyme:
(-)-Fucose, 6-Deoxy-L-Galactose, (2S,3R,4R,5S)-2,3,4,5-Tetrahydroxyhexanal
A16789.03
A16789.06
A16789.03EA
45.7
EUR
A16789.03
A16789.06
L(-)-Fucose 98%
L(-)-Fucose
Purity: 99%
CAS No.: 2438-80-4
Molecular Formula: C6H12O5
Formula Weight: 164.16
Melting Point: 139-142°
Beilstein Reference: 1422661
Merck Reference: 14,4279
Optical Rotation: -75° (c=10 in water)
MDL No.: MFCD00069812
Formel:
C₆H₁₂O₅ Molecular Weight: 164,16 g/mol Schmelzpunkt: 139…142 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00069812 CAS-Nummer: 2438-80-4 EINECS: 219-452-7 Merck Index: 14,4279 |