4-Amino-2,5-dimethylphenol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Amino-2,5-dimethylphenol

M-Clarity™ Qualitätsstufe = MQ100

126497-10G 126497-5G
SIAL126497-10GEA 142 EUR
SIAL126497-10G SIAL126497-5G
4-Amino-2,5-dimethylphenol, Sigma-Aldrich®
4-Amino-2,5-xylenol
Formel: C₈H₁₁NO
Molecular Weight: 137,1800 g/mol
MDL: MFCD00007873
CAS-Nummer: 3096-71-7
EINECS: 221-449-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR