2-Fluor-5-(trifluormethyl)benzophenon

Lieferant: FLUOROCHEM

Synonyme: (2-Fluoro-5-(trifluoromethyl)phenyl)(phenyl)methanone

006397-1G 006397-5G 006397-10G
FLUO006397-1GEA 36.9 EUR
FLUO006397-1G FLUO006397-5G FLUO006397-10G
2-Fluor-5-(trifluormethyl)benzophenon
2-Fluor-5-(trifluormethyl)benzophenon
Formel: C₁₄H₈F₄O
MDL: MFCD00061254
CAS-Nummer: 199292-40-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR