2-Fluor-4-(trifluormethyl)benzophenon

Lieferant: FLUOROCHEM

006396-1G 006396-5G 006396-10G
FLUO006396-1GEA 121 EUR
FLUO006396-1G FLUO006396-5G FLUO006396-10G
2-Fluor-4-(trifluormethyl)benzophenon
2-Fluor-4-(trifluormethyl)benzophenon
Formel: C₁₄H₈F₄O
Molecular Weight: 268,21 g/mol
MDL: MFCD00061253
CAS-Nummer: 207974-08-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR