4-(Methylsulfanyl)-1-butanol ≥99%

Lieferant: Thermo Fisher Scientific

B22673.03 B22673.06 B22673.14
B22673.03EA 31.9 EUR
B22673.03 B22673.06 B22673.14
4-(Methylsulfanyl)-1-butanol ≥99%
4-(Methylthio)butanol

Purity: 99% CAS No.: 20582-85-8 Molecular Formula: C5H12OS Formula Weight: 120.21 Flash Point: 101°(215°F) Refractive Index: 1.4870 Odour: Unpleasant Boiling Point: 95-98°/20mm MDL No.: MFCD00002973

Formel: C₅H₁₂OS
Molecular Weight: 120.21 g/mol
Siedepunkt: 95…98 °C (20 mmHg)
Dichte: 0.999
Flammpunkt: 101 °C (215 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002973
CAS-Nummer: 20582-85-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR